Tisk…

8-Bromo-1,6-naphthyridin-5-amine 97%

Dodavatel: Apollo Scientific

APOSOR53039-250MGEA 5000 CZK
APOSOR53039-250MG
8-Bromo-1,6-naphthyridin-5-amine 97%
8-Bromo-1,6-naphthyridin-5-amine
Vzorec: C₈H₆BrN₃
MW: 224,057 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22396116
CAS číslo: 1820686-20-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR