3-Amino-5-(tert-butyl)thiophene-2-carboxamide 98%
Dodavatel: Apollo Scientific
|
Varování
|
APOSOR5325-100MGEA
457
CZK
APOSOR5325-100MG
3-Amino-5-(tert-butyl)thiophene-2-carboxamide 98%
3-Amino-5-(tert-butyl)thiophene-2-carboxamide
Vzorec:
C₉H₁₄N₂OS MW: 198,285 g/mol Bod tání: 146…148 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00068165 CAS číslo: 175137-04-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...