Tisk…
(S)-Methyl N-tert-butoxycarbonyl-3-(4-biphenylyl)-2-aminopropionate 98%
Dodavatel: Apollo Scientific
APOSOR5402-5GEA 72740 CZK
APOSOR5402-5G
(S)-Methyl N-tert-butoxycarbonyl-3-(4-biphenylyl)-2-aminopropionate 98%
(S)-Methyl N-tert-butoxycarbonyl-3-(4-biphenylyl)-2-aminopropionate
Vzorec: C₂₁H₂₅NO₄
MW: 355,427 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01075194
CAS číslo: 137255-86-8