Tisk…

2-Methyl-N-phenylaniline 98%

Dodavatel: Apollo Scientific

APOSOR54435-1GEA 5000 CZK
APOSOR54435-1G
2-Methyl-N-phenylaniline 98%
2-Methyl-N-phenylaniline
Vzorec: C₁₃H₁₃N
MW: 183,249 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01318642
CAS číslo: 1205-39-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR