Tisk…

2-Hydroxy-4-cyclopropylbenzaldehyde

Dodavatel: Apollo Scientific

Varování

APOSOR54662-250MGEA 7280 CZK
APOSOR54662-250MG
2-Hydroxy-4-cyclopropylbenzaldehyde
2-Hydroxy-4-cyclopropylbenzaldehyde
Vzorec: C₁₀H₁₀O₂
MW: 162,185 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31726536
CAS číslo: 2387695-91-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR