Tisk…

2-Amino-2-(3,4-dichlorophenyl)ethanol 97%

Dodavatel: Apollo Scientific

APOSOR55015-250MGEA 6850 CZK
APOSOR55015-250MG
2-Amino-2-(3,4-dichlorophenyl)ethanol 97%
2-Amino-2-(3,4-dichlorophenyl)ethanol
Vzorec: C₈H₉Cl₂NO
MW: 206,069 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12827189
CAS číslo: 188586-38-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR