Tisk…

2-(2-Bromothiazol-4-yl)acetic acid

Dodavatel: Apollo Scientific

APOSOR55097-100MGEA 8200 CZK
APOSOR55097-100MG
2-(2-Bromothiazol-4-yl)acetic acid
2-(2-Bromothiazol-4-yl)acetic acid
Vzorec: C₅H₄BrNO₂S
MW: 222,06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18410768
CAS číslo: 62557-07-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR