1-[2-(Azetidin-1-yl)ethyl]-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Dodavatel: Apollo Scientific
|
Varování
|
APOSOR55283-250MGEA
13060
CZK
APOSOR55283-250MG
1-[2-(Azetidin-1-yl)ethyl]-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
1-[2-(Azetidin-1-yl)ethyl]-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Vzorec:
C₁₄H₂₄BN₃O₂ MW: 277,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27987628 CAS číslo: 1422126-12-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...