Tisk…

3-Bromo-7-iodoquinoline

Dodavatel: Apollo Scientific

Varování

APOSOR55324-250MGEA 4570 CZK
APOSOR55324-250MG APOSOR55324-1G
3-Bromo-7-iodoquinoline
3-Bromo-7-iodoquinoline
Vzorec: C₉H₅BrIN
MW: 333,951 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28679234
CAS číslo: 1354223-46-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR