2-Chloro-5-(dimethoxymethyl)aniline 95%
Dodavatel: Apollo Scientific
APOSOR55365-5GEA
33400
CZK
APOSOR55365-5G
2-Chloro-5-(dimethoxymethyl)aniline 95%
2-Chloro-5-(dimethoxymethyl)aniline
Vzorec:
C₉H₁₂ClNO₂ MW: 201,65 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31614277 CAS číslo: 2244087-36-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...