Tisk…

4-Chloro-3-(dimethoxymethyl)aniline 95%

Dodavatel: Apollo Scientific

APOSOR55369-5GEA 33400 CZK
APOSOR55369-5G
4-Chloro-3-(dimethoxymethyl)aniline 95%
4-Chloro-3-(dimethoxymethyl)aniline
Vzorec: C₉H₁₂ClNO₂
MW: 201,65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31614279
CAS číslo: 2244085-05-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR