3-(Dimethoxymethyl)-4-hydroxyaniline 95%
Dodavatel: Apollo Scientific
APOSOR55373-5GEA
33400
CZK
APOSOR55373-5G
3-(Dimethoxymethyl)-4-hydroxyaniline 95%
3-(Dimethoxymethyl)-4-hydroxyaniline
Vzorec:
C₉H₁₃NO₃ MW: 183,204 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20718402 CAS číslo: 197707-52-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...