Tisk…

3-Bromo- N,N-dimethyl-4-methoxyaniline 95%

Dodavatel: Apollo Scientific

APOSOR55523-1GEA 20700 CZK
APOSOR55523-1G
3-Bromo- N,N-dimethyl-4-methoxyaniline 95%
3-Bromo- N,N-dimethyl-4-methoxyaniline
Vzorec: C₉H₁₂BrNO
MW: 230,102 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16315291
CAS číslo: 102236-13-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR