5-Amino-2-methoxystyrene 95%
Dodavatel: Apollo Scientific
APOSOR55528-1GEA
33400
CZK
APOSOR55528-1G
5-Amino-2-methoxystyrene 95%
5-Amino-2-methoxystyrene
Vzorec:
C₉H₁₁NO MW: 149,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31629943 CAS číslo: 2244088-80-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...