Tisk…

(R)-3-Cyclopropyl-2-hydroxypropanoic acid 95%

Dodavatel: Apollo Scientific

Varování

APOSOR55546-250MGEA 28570 CZK
APOSOR55546-250MG
(R)-3-Cyclopropyl-2-hydroxypropanoic acid 95%
(R)-3-Cyclopropyl-2-hydroxypropanoic acid
Vzorec: C₆H₁₀O₃
MW: 130,142 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17171014
CAS číslo: 174265-97-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR