Tisk…

(2-Bromophenyl)(dimethylamino)acetonitrile

Dodavatel: Apollo Scientific

Varování

APOSOR59401-1GEA 5000 CZK
APOSOR59401-1G
(2-Bromophenyl)(dimethylamino)acetonitrile
(2-Bromophenyl)(dimethylamino)acetonitrile
Vzorec: C₁₀H₁₁BrN₂
MW: 239,112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878365
CAS číslo: 299215-38-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR