Tisk…

Ethyl thiomorpholine-3-carboxylate hydrochloride

Dodavatel: Apollo Scientific

Varování

APOSOR59472-1GEA 3770 CZK
APOSOR59472-1G
Ethyl thiomorpholine-3-carboxylate hydrochloride
Ethyl thiomorpholine-3-carboxylate hydrochloride
Vzorec: C₇H₁₃NO₂S·ClH
MW: 211,71 g/mol
Bod tání: 133…136 °C
Storage Temperature: Ambient
MDL Number: MFCD00107968
CAS číslo: 159381-07-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR