Tisk…

2-Amino-4-(3-nitrophenyl)pyrimidine 97%

Dodavatel: Apollo Scientific

APOSOR60125-250MGEA 2450 CZK
APOSOR60125-250MG
2-Amino-4-(3-nitrophenyl)pyrimidine 97%
2-Amino-4-(3-nitrophenyl)pyrimidine
Vzorec: C₁₀H₈N₄O₂
MW: 216,196 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00665910
CAS číslo: 240136-69-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR