Tisk…

2-(Aminomethyl)-5-bromo-4-methylpyridine hydrochloride

Dodavatel: Apollo Scientific

Varování

APOSOR60141-250MGEA 4340 CZK
APOSOR60141-250MG
2-(Aminomethyl)-5-bromo-4-methylpyridine hydrochloride
2-(Aminomethyl)-5-bromo-4-methylpyridine hydrochloride
Vzorec: C₇H₉BrN₂·ClH
MW: 237,525 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17214201
CAS číslo: 1257535-47-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR