Tisk…

4-(Pyridin-3-ylsulphonyl)morpholine 98%

Dodavatel: Apollo Scientific

APOSOR6092-1GEA 2740 CZK
APOSOR6092-1G
4-(Pyridin-3-ylsulphonyl)morpholine 98%
4-(Pyridin-3-ylsulphonyl)morpholine
Vzorec: C₉H₁₂N₂O₃S
MW: 228,268 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00090297
CAS číslo: 26103-48-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR