2-(Benzhydrylthio)acetamide 98%
Dodavatel: Apollo Scientific
APOSOR61208-25GEA
3280
CZK
APOSOR61208-25G
2-(Benzhydrylthio)acetamide 98%
2-(Benzhydrylthio)acetamide
Vzorec:
C₁₅H₁₅NOS MW: 257,351 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00876107 CAS číslo: 68524-30-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...