Tisk…

2-Chloro-4-isopropoxy-5-nitrotoluene 97%

Dodavatel: Apollo Scientific
Varování

APOSOR61251-250MGEA 4400 CZK
APOSOR61251-250MG
2-Chloro-4-isopropoxy-5-nitrotoluene 97%
2-Chloro-4-isopropoxy-5-nitrotoluene
Vzorec: C₁₀H₁₂ClNO₃
MW: 229,66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11499001
CAS číslo: 1032903-50-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR