Tisk…

(3S)-3-(Isobutylamino)piperidine, N1-BOC protected

Dodavatel: Apollo Scientific

APOSOR62009-1GEA 17820 CZK
APOSOR62009-1G
(3S)-3-(Isobutylamino)piperidine, N1-BOC protected
(3S)-3-(Isobutylamino)piperidine, N1-BOC protected
Vzorec: C₁₄H₂₈N₂O₂
MW: 256,384 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19381684
CAS číslo: 1027346-21-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR