Tisk…

4-[(Piperidin-1-yl)methyl]benzyl alcohol

Dodavatel: Apollo Scientific

Nebezpečí

APOSOR6752-1GEA 3340 CZK
APOSOR6752-1G
4-[(Piperidin-1-yl)methyl]benzyl alcohol
4-[(Piperidin-1-yl)methyl]benzyl alcohol
Vzorec: C₁₃H₁₉NO
MW: 205,296 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07772863
CAS číslo: 91271-62-4
UN: 3259
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR