Tisk…

2-Amino-3'-chlorobiphenyl hydrochloride

Dodavatel: Apollo Scientific

Varování

APOSOR7337-50MGEA 1040 CZK
APOSOR7337-50MG
2-Amino-3'-chlorobiphenyl hydrochloride
2-Amino-3'-chlorobiphenyl hydrochloride
Vzorec: C₁₂H₁₀ClN·ClH
MW: 240,128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06739421
CAS číslo: 1172032-93-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR