Tisk…

3-Acetyl-2'-chlorobiphenyl

Dodavatel: Apollo Scientific

Varování

APOSOR7367-250MGEA 3440 CZK
APOSOR7367-250MG
3-Acetyl-2'-chlorobiphenyl
3-Acetyl-2'-chlorobiphenyl
Vzorec: C₁₄H₁₁ClO
MW: 230,69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039058
CAS číslo: 675596-35-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR