Tisk…

2-(Thien-3-yl)benzaldehyde

Dodavatel: Apollo Scientific

Varování

APOSOR7590-1GEA 8470 CZK
APOSOR7590-1G
2-(Thien-3-yl)benzaldehyde
2-(Thien-3-yl)benzaldehyde
Vzorec: C₁₁H₈OS
MW: 188,246 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039149
CAS číslo: 99902-03-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR