Tisk…

2-(Benzyloxycarbonylamino)glutarimide 98%

Dodavatel: Apollo Scientific

Varování

APOSOR8165-1GEA 7870 CZK
APOSOR8165-1G
2-(Benzyloxycarbonylamino)glutarimide 98%
2-(Benzyloxycarbonylamino)glutarimide
Vzorec: C₁₃H₁₄N₂O₄
MW: 262,261 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01075195
CAS číslo: 24666-55-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR