Benzyl-α-D-mannopyranoside
Dodavatel: Apollo Scientific
APOSOR8200T-1GEA
3740
CZK
APOSOR8200T-1G
Benzyl-α-D-mannopyranoside
Benzyl-α-D-mannopyranoside
Vzorec:
C₁₃H₁₈O₆ MW: 270,278 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00153924 CAS číslo: 15548-45-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...