Tisk…

1-(2,6-Dimethylphenyl)-1H-pyrrole-3-carbaldehyde 95%

Dodavatel: Apollo Scientific

APOSOR902470-1GEA 9960 CZK
APOSOR902470-1G
1-(2,6-Dimethylphenyl)-1H-pyrrole-3-carbaldehyde 95%
1-(2,6-Dimethylphenyl)-1H-pyrrole-3-carbaldehyde
Vzorec: C₁₃H₁₃NO
MW: 199,248 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06653456
CAS číslo: 132688-31-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR