Tisk…

4-Amino-2-chloro-5-methylpyridin-1-ium-1-olate 95%

Dodavatel: Apollo Scientific

APOSOR902697-1GEA 18020 CZK
APOSOR902697-1G
4-Amino-2-chloro-5-methylpyridin-1-ium-1-olate 95%
4-Amino-2-chloro-5-methylpyridin-1-ium-1-olate
Vzorec: C₆H₇ClN₂O
MW: 158,586 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22192434
CAS číslo: 1373232-42-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR