Tisk…

4-[(3aS,4R,9bR)-6-Ethoxy-3H,3aH,4H,5H,9bH-cyclopenta[c]quinolin-4-yl]benzoic acid

Dodavatel: Apollo Scientific

Varování

APOSOR902994-5GEA 13260 CZK
APOSOR902994-5G
4-[(3aS,4R,9bR)-6-Ethoxy-3H,3aH,4H,5H,9bH-cyclopenta[c]quinolin-4-yl]benzoic acid
4-[(3aS,4R,9bR)-6-Ethoxy-3H,3aH,4H,5H,9bH-cyclopenta[c]quinolin-4-yl]benzoic acid
Vzorec: C₂₁H₂₁NO₃
MW: 335,396 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07403664
CAS číslo: 956189-58-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR