Tisk…

5-Chloro-2-methylphenyl isocyanide 95%

Dodavatel: Apollo Scientific

APOSOR903321-1GEA 18020 CZK
APOSOR903321-1G
5-Chloro-2-methylphenyl isocyanide 95%
5-Chloro-2-methylphenyl isocyanide
Vzorec: C₈H₆ClN
MW: 151,593 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02664603
CAS číslo: 602262-01-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR