tert-Butyl N-(1-cyano-1-methylethyl)carbamate 95%
Dodavatel: Apollo Scientific
APOSOR903352-1GEA
15340
CZK
APOSOR903352-1G
tert-Butyl N-(1-cyano-1-methylethyl)carbamate 95%
tert-Butyl N-(1-cyano-1-methylethyl)carbamate
Vzorec:
C₉H₁₆N₂O₂ MW: 184,236 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12791990 CAS číslo: 133117-97-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...