Tisk…

(4-Methoxy-2,5-dimethylphenyl)acetic acid 95%

Dodavatel: Apollo Scientific

APOSOR903491-1GEA 18020 CZK
APOSOR903491-1G
(4-Methoxy-2,5-dimethylphenyl)acetic acid 95%
(4-Methoxy-2,5-dimethylphenyl)acetic acid
Vzorec: C₁₁H₁₄O₃
MW: 194,227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06208110
CAS číslo: 104216-78-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR