Tisk…

1-Isocyano-3,5-dimethyl-benzene 95%

Dodavatel: Apollo Scientific

APOSOR903616-1GEA 9960 CZK
APOSOR903616-1G
1-Isocyano-3,5-dimethyl-benzene 95%
1-Isocyano-3,5-dimethyl-benzene
Vzorec: C₉H₉N
MW: 131,174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02664617
CAS číslo: 20600-56-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR