Tisk…

2-(1-Methyl-2-piperidinyl)azepane 95%

Dodavatel: Apollo Scientific

APOSOR904486-1GEA 18020 CZK
APOSOR904486-1G
2-(1-Methyl-2-piperidinyl)azepane 95%
2-(1-Methyl-2-piperidinyl)azepane
Vzorec: C₁₂H₂₄N₂
MW: 196,332 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02663725
CAS číslo: 383129-17-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR