Tisk…

2-(1-Methyl-4-piperidinyl)azepane 95%

Dodavatel: Apollo Scientific

APOSOR904562-1GEA 9960 CZK
APOSOR904562-1G
2-(1-Methyl-4-piperidinyl)azepane 95%
2-(1-Methyl-4-piperidinyl)azepane
Vzorec: C₁₂H₂₄N₂
MW: 196,332 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05190303
CAS číslo: 527674-00-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR