Tisk…

1-(2,6-Dimethylphenyl)-1H-pyrrole 95%

Dodavatel: Apollo Scientific

APOSOR904686-1GEA 9960 CZK
APOSOR904686-1G
1-(2,6-Dimethylphenyl)-1H-pyrrole 95%
1-(2,6-Dimethylphenyl)-1H-pyrrole
Vzorec: C₁₂H₁₃N
MW: 171,238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03195931
CAS číslo: 15898-23-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR