Tisk…

2-Cyclopentylpiperidine hydrochloride 95%

Dodavatel: Apollo Scientific

APOSOR905476-1GEA 18020 CZK
APOSOR905476-1G
2-Cyclopentylpiperidine hydrochloride 95%
2-Cyclopentylpiperidine hydrochloride
Vzorec: C₁₀H₁₉N·ClH
MW: 189,725 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08277055
CAS číslo: 1177350-52-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR