Tisk…

4-Bromo-2-methylphenyl isocyanide 95%

Dodavatel: Apollo Scientific

APOSOR905490-1GEA 9960 CZK
APOSOR905490-1G
4-Bromo-2-methylphenyl isocyanide 95%
4-Bromo-2-methylphenyl isocyanide
Vzorec: C₈H₆BrN
MW: 196,044 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07389710
CAS číslo: 1258794-26-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR