Tisk…

2-Amino-2-methyl-butyric acid methyl ester hydrochloride 96%

Dodavatel: Apollo Scientific

APOSOR905498-1GEA 10350 CZK
APOSOR905498-1G
2-Amino-2-methyl-butyric acid methyl ester hydrochloride 96%
2-Amino-2-methyl-butyric acid methyl ester hydrochloride
Vzorec: C₆H₁₃NO₂·ClH
MW: 167,634 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06737779
CAS číslo: 156032-14-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR