Tisk…

3-Methyl-2-(1H-pyrrol-1-yl)pentanoic acid 95%

Dodavatel: Apollo Scientific

APOSOR905517-1GEA 18020 CZK
APOSOR905517-1G
3-Methyl-2-(1H-pyrrol-1-yl)pentanoic acid 95%
3-Methyl-2-(1H-pyrrol-1-yl)pentanoic acid
Vzorec: C₁₀H₁₅NO₂
MW: 181,232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08752782
CAS číslo: 63751-73-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR