Tisk…

2-(2-Methylbenzyl)piperidine 95%

Dodavatel: Apollo Scientific

APOSOR905544-1GEA 18020 CZK
APOSOR905544-1G
2-(2-Methylbenzyl)piperidine 95%
2-(2-Methylbenzyl)piperidine
Vzorec: C₁₃H₁₉N
MW: 189,297 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02663587
CAS číslo: 383128-20-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR