3-Methyl-2-(2-pyrrolidinylmethyl)pyridine 95%
Dodavatel: Apollo Scientific
APOSOR905546-1GEA
18020
CZK
APOSOR905546-1G
3-Methyl-2-(2-pyrrolidinylmethyl)pyridine 95%
3-Methyl-2-(2-pyrrolidinylmethyl)pyridine
Vzorec:
C₁₁H₁₆N₂ MW: 176,258 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06653323 CAS číslo: 881040-09-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...