Tisk…

2-Cyclohexylazepane 95%

Dodavatel: Apollo Scientific

APOSOR906187-250MGEA 10350 CZK
APOSOR906187-250MG APOSOR906187-1G
2-Cyclohexylazepane 95%
2-Cyclohexylazepane
Vzorec: C₁₂H₂₃N
MW: 181,318 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02663723
CAS číslo: 383129-16-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR