4-(1-Isocyanoethyl)pyridine 95%
Dodavatel: Apollo Scientific
APOSOR906421-1GEA
18020
CZK
APOSOR906421-1G
4-(1-Isocyanoethyl)pyridine 95%
4-(1-Isocyanoethyl)pyridine
Vzorec:
C₈H₈N₂ MW: 132,163 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05222983 CAS číslo: 87869-17-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...