Ethyl 2-isocyanopropanoate 95%
Dodavatel: Apollo Scientific
APOSOR906638-100MGEA
27180
CZK
APOSOR906638-100MG
Ethyl 2-isocyanopropanoate 95%
Ethyl 2-isocyanopropanoate
Vzorec:
C₆H₉NO₂ MW: 127,141 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02664649 CAS číslo: 33140-27-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...