Tisk…

3-Chloro-4-methylphenylisocyanide 95%

Dodavatel: Apollo Scientific

APOSOR906836-1GEA 20500 CZK
APOSOR906836-1G APOSOR906836-250MG
3-Chloro-4-methylphenylisocyanide 95%
3-Chloro-4-methylphenylisocyanide
Vzorec: C₈H₆ClN
MW: 151,593 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02664604
CAS číslo: 112675-35-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR