Tisk…

3-Chlorophthalide 95%

Dodavatel: Apollo Scientific

APOSOR906923-5GEA 27540 CZK
APOSOR906923-5G APOSOR906923-1G
3-Chlorophthalide 95%
3-Chlorophthalide
Vzorec: C₈H₅ClO₂
MW: 168,577 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011556
CAS číslo: 6295-21-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR